siRNA Details
| Gene Symbol: CCDC142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032779.3 | NP_116168.3 | 9 | 2620 | ||
| GenBank mRNA | AK027303.1 | - | 3 | 818 | ||
| AK075543.1 | - | 9 | 2386 | |||
| BC117269.1 | - | 9 | 2338 | 40125820 | ||
| BC143398.1 | - | 9 | 2338 | 9051905 | ||
| BC143399.1 | - | 8 | 2451 | |||

CCDC142
Genomic Map