siRNA Details
| Gene Symbol: SLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003063.2 | NP_003054.1 | 2 | 666 | ||
| GenBank mRNA | BC005261.1 | - | 2 | 661 | 3934241 | |
| BC094685.1 | - | 2 | 648 | 6203008 | ||
| F30315.1 | - | 1 | 280 | |||
| U96094.1 | - | 2 | 660 | |||

SLN
Genomic Map