siRNA Details
| Gene Symbol: MCRIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138418.2 | NP_612427.2 | 905 | |||
| GenBank mRNA | BC001912.1 | - | 905 | 3546959 | ||
| BC007346.2 | - | 697 | ||||
| BC111391.1 | - | 808 | 6457301 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MCRIP2| Gene Symbol: MCRIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138418.2 | NP_612427.2 | 905 | |||
| GenBank mRNA | BC001912.1 | - | 905 | 3546959 | ||
| BC007346.2 | - | 697 | ||||
| BC111391.1 | - | 808 | 6457301 | |||