siRNA Details
Gene Symbol: CCDC142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032779.3 | NP_116168.3 | 9 | 2693 | ||
GenBank mRNA | AK027303.1 | - | 3 | 891 | ||
AK075543.1 | - | 9 | 2459 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CCDC142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032779.3 | NP_116168.3 | 9 | 2693 | ||
GenBank mRNA | AK027303.1 | - | 3 | 891 | ||
AK075543.1 | - | 9 | 2459 |