siRNA Details
| Gene Symbol: GPR62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080865.3 | NP_543141.3 | 1 | 2182 | ||
| GenBank mRNA | BC026357.1 | - | 1 | 2115 | 4812283 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR62| Gene Symbol: GPR62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080865.3 | NP_543141.3 | 1 | 2182 | ||
| GenBank mRNA | BC026357.1 | - | 1 | 2115 | 4812283 | |
Set Membership: |
|