siRNA Details
| Gene Symbol: LOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000427.2 | NP_000418.2 | 2 | 1031 | ||
| GenBank mRNA | BC034690.1 | - | 1 | 1022 | 4752776 | |
| BC108290.1 | - | 2 | 1043 | 4754528 | ||
| M61120.1 | - | 1 | 1022 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOR| Gene Symbol: LOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000427.2 | NP_000418.2 | 2 | 1031 | ||
| GenBank mRNA | BC034690.1 | - | 1 | 1022 | 4752776 | |
| BC108290.1 | - | 2 | 1043 | 4754528 | ||
| M61120.1 | - | 1 | 1022 | |||