siRNA Details
| Gene Symbol: CHRM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000738.2 | NP_000729.2 | 2 | 482 | ||
| XM_011544742.2 | XP_011543044.1 | 2 | 183 | |||
| GenBank mRNA | AF385587.1 | - | 1 | 91 | ||
| AF498915.1 | - | 1 | 81 | |||
| AK312374.1 | - | 2 | 243 | |||
| BC007740.2 | - | 2 | 203 | 4128514 | ||
| BC022984.1 | - | 2 | 482 | 4931283 | ||
| BT007166.1 | - | 1 | 81 | |||
More Information
Set Membership: |
|

CHRM1
Genomic Map