siRNA Details
| Gene Symbol: GAGE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102272.1 | 4, 5 | 459 | |||
| GenBank mRNA | BC036094.1 | - | 4, 5 | 447 | 5296612 | |
| U19142.1 | - | 4, 5 | 381 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GAGE1| Gene Symbol: GAGE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102272.1 | 4, 5 | 459 | |||
| GenBank mRNA | BC036094.1 | - | 4, 5 | 447 | 5296612 | |
| U19142.1 | - | 4, 5 | 381 | |||