siRNA Details
| Gene Symbol: OR1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002548.2 | NP_002539.2 | 1 | 418 | ||
| GenBank mRNA | BC069552.1 | - | 1 | 418 | 7262253 | |
| BC106735.1 | - | 1 | 418 | 40015766 | ||
| BC106736.1 | - | 1 | 418 | 40015767 | ||

OR1D2
Genomic Map