siRNA Details
| Gene Symbol: GAGE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102272.1 | 5 | 488 | |||
| GenBank mRNA | BC036094.1 | - | 5 | 476 | 5296612 | |
| U19142.1 | - | 5 | 410 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GAGE1| Gene Symbol: GAGE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102272.1 | 5 | 488 | |||
| GenBank mRNA | BC036094.1 | - | 5 | 476 | 5296612 | |
| U19142.1 | - | 5 | 410 | |||