siRNA Details
| Gene Symbol: CXCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001511.3 | NP_001502.1 | 3 | 337 | ||
| NR_046035.1 | 3 | 360 | ||||
| GenBank mRNA | BC011976.1 | - | 3 | 325 | 3856841 | |
| BG121608.1 | - | 3 | 344 | |||
| BM554448.1 | - | 3 | 350 | |||
| BT006880.1 | - | 3 | 258 | |||
| CB112830.1 | - | 3 | 338 | |||
| J03561.1 | - | 3 | 311 | |||
| X12510.1 | - | 3 | 275 | |||

CXCL1
Genomic Map