siRNA Details
| Gene Symbol: SNAPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003083.3 | NP_003074.1 | 2 | 294 | ||
| NR_030717.1 | 2 | 191 | ||||
| GenBank mRNA | AK222926.1 | - | 2 | 265 | ||
| AK314878.1 | - | 2 | 279 | |||
| BC011868.2 | - | 2 | 249 | 4545178 | ||
| BP234322.1 | - | 2 | 190 | |||
| BX647416.1 | - | 2 | 158 | |||
| CF454397.1 | - | 2 | 294 | |||
| U44755.1 | - | 2 | 278 | |||
| U44898.1 | - | 2 | 260 | |||

SNAPC2
Genomic Map