siRNA Details
| Gene Symbol: TYROBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173514.1 | NP_001166985.1 | 4 | 399 | ||
| NM_001173515.1 | NP_001166986.1 | 4 | 396 | |||
| NM_003332.3 | NP_003323.1 | 5 | 432 | |||
| NM_198125.2 | NP_937758.1 | 5 | 429 | |||
| NR_033390.1 | 4 | 392 | ||||
| GenBank mRNA | AF019562.1 | - | 5 | 418 | ||
| AK290385.1 | - | 5 | 415 | |||
| BC011175.1 | - | 5 | 407 | 4150408 | ||
| BI908458.1 | - | 4 | 363 | |||
| BP295666.1 | - | 4 | 365 | |||
| BP302545.1 | - | 5 | 432 | |||
| BP305672.1 | - | 4 | 383 | |||

TYROBP
Genomic Map