siRNA Details
| Gene Symbol: UBE2D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204880.1 | NP_001191809.1 | 3 | 344 | ||
| NM_003338.4 | NP_003329.1 | 4 | 376 | |||
| XM_017016609.1 | XP_016872098.1 | 4 | 275 | |||
| GenBank mRNA | AF257220.1 | - | 4 | 299 | ||
| AJ272367.1 | - | 4 | 299 | |||
| AK291901.1 | - | 4 | 347 | |||
| BC005980.1 | - | 4 | 347 | 4106710 | ||
| BC015997.1 | - | 4 | 351 | 4305900 | ||
| BI598342.1 | - | 4 | 380 | |||
| BT007041.1 | - | 4 | 153 | |||
| DA618122.1 | - | 3 | 317 | |||
| DA677507.1 | - | 4 | 376 | |||
| X78140.1 | - | 4 | 168 | |||

UBE2D1
Genomic Map