siRNA Details
| Gene Symbol: HIST1H3H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003536.2 | NP_003527.1 | 1 | 209 | ||
| GenBank mRNA | AK311991.1 | - | 1 | 222 | ||
| BC007518.2 | - | 1 | 337 | |||
| BC096128.3 | - | 1 | 199 | 40002508 | ||
| BC096129.3 | - | 1 | 199 | 40002509 | ||
| BC096130.3 | - | 1 | 199 | 40002510 | ||
| BC143046.1 | - | 1 | 199 | 40112088 | ||
| BC148243.1 | - | 1 | 199 | 40082529 | ||
| BC148250.1 | - | 1 | 199 | 40002516 | ||

HIST1H3H
Genomic Map