siRNA Details
Gene Symbol: PITX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005029.3 | NP_005020.1 | 2, 3 | 273 | ||
GenBank mRNA | AF041339.1 | - | 1, 2 | 122 | ||
BC011642.2 | - | 2, 3 | 272 | 4131442 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PITX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005029.3 | NP_005020.1 | 2, 3 | 273 | ||
GenBank mRNA | AF041339.1 | - | 1, 2 | 122 | ||
BC011642.2 | - | 2, 3 | 272 | 4131442 |