siRNA Details
| Gene Symbol: MAGEA12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166386.3 | NP_001159858.1 | 3 | 625 | ||
| NM_001166387.3 | NP_001159859.1 | 3 | 533 | |||
| NM_005367.6 | NP_005358.2 | 2 | 427 | |||
| GenBank mRNA | AK314168.1 | - | 3 | 625 | ||
| BC003408.2 | - | 2 | 415 | 3450217 | ||
| BC098146.1 | - | 3 | 205 | |||
| BQ436888.1 | - | 2 | 427 | |||
| BT007108.1 | - | 1 | 271 | |||
| CR541775.1 | - | 1 | 271 | |||

MAGEA12
Genomic Map