siRNA Details
| Gene Symbol: CLEC3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003278.2 | NP_003269.2 | 1, 2 | 213 | ||
| GenBank mRNA | AK312093.1 | - | 1, 2 | 221 | ||
| BC011024.1 | - | 1, 2 | 178 | 4042921 | ||
| BI837909.1 | - | 1, 2 | 297 | |||
| CD616729.1 | - | 1, 2 | 187 | |||
| CR541981.1 | - | 1, 2 | 117 | |||
| CR542009.1 | - | 1, 2 | 117 | |||
| X64559.1 | - | 1, 2 | 210 | |||
 Back To Top
 Back To Top
 CLEC3B
CLEC3B Genomic Map
Genomic Map Back To Top
 Back To Top