siRNA Details
| Gene Symbol: TMEM187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003492.2 | NP_003483.1 | 2 | 1166 | ||
| XM_011531199.2 | XP_011529501.1 | 2 | 1181 | |||
| GenBank mRNA | AK314938.1 | - | 2 | 946 | ||
| BC008203.1 | - | 2 | 914 | 3868838 | ||
| BC017026.1 | - | 2 | 901 | 3847234 | ||
| CR457047.1 | - | 1 | 632 | |||
| X92475.1 | - | 2 | 916 | |||

TMEM187
Genomic Map