siRNA Details
Gene Symbol: SUPT6H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320755.1 | NP_001307684.1 | 4 | 406 | ||
NM_003170.4 | NP_003161.2 | 3 | 304 | |||
XM_017024978.1 | XP_016880467.1 | 5 | 465 | |||
XM_017024979.1 | XP_016880468.1 | 3 | 542 | |||
XM_017024980.1 | XP_016880469.1 | 2 | 236 | |||
GenBank mRNA | AL833607.1 | - | 4 | 407 | ||
BC136522.1 | - | 3 | 216 | 9020512 | ||
BC136524.1 | - | 3 | 216 | 9020514 | ||
BC150268.1 | - | 3 | 218 | 9007230 | ||
D79984.1 | - | 3 | 218 |
