siRNA Details
| Gene Symbol: FAM127A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001078171.1 | NP_001071639.1 | 1 | 1090 | ||
| GenBank mRNA | AF038168.1 | - | 1 | 1026 | ||
| AF052096.1 | - | 1 | 1026 | |||
| BC002385.2 | - | 1 | 1020 | 2961665 | ||
| BC002410.2 | - | 1 | 1020 | 2961641 | ||
| Y13374.1 | - | 1 | 1035 | |||

FAM127A
Genomic Map