siRNA Details
| Gene Symbol: RBM8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005105.4 | NP_005096.1 | 6 | 676 | ||
| GenBank mRNA | AF127761.1 | - | 6 | 578 | ||
| AF161463.1 | - | 6 | 590 | |||
| AF182415.1 | - | 6 | 595 | |||
| AF198620.1 | - | 6 | 597 | |||
| AF231511.1 | - | 6 | 583 | |||
| AK075009.1 | - | 6 | 597 | |||
| BC017088.2 | - | 6 | 638 | 3850355 | ||
| BC071577.1 | - | 6 | 624 | |||

RBM8A
Genomic Map