siRNA Details
| Gene Symbol: DNAJC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001307980.1 | NP_001294909.1 | 3 | 776 | ||
| NM_001307981.1 | NP_001294910.1 | 3 | 776 | |||
| NM_005528.3 | NP_005519.2 | 4 | 815 | |||
| GenBank mRNA | AF012106.1 | - | 1 | 990 | ||
| AF036874.1 | - | 3 | 422 | |||
| BC032137.2 | - | 3 | 450 | |||
| BC044584.1 | - | 4 | 818 | 4811478 | ||
| BC051008.1 | - | 3 | 298 | |||
| BC053947.1 | - | 4 | 818 | |||
| BC057849.1 | - | 3 | 648 | 30349314 | ||

DNAJC4
Genomic Map