siRNA Details
| Gene Symbol: NKG7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005601.3 | NP_005592.1 | 1 | 250 | ||
| XM_005258955.3 | XP_005259012.1 | 1 | 232 | |||
| XM_006723228.3 | XP_006723291.1 | 1 | 232 | |||
| GenBank mRNA | BC015759.1 | - | 1 | 179 | 4849734 | |
| S69115.1 | - | 1 | 259 | |||

NKG7
Genomic Map