siRNA Details
| Gene Symbol: CAPN15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005632.2 | NP_005623.1 | 4 | 1327 | ||
| XM_011522620.2 | XP_011520922.1 | 5 | 4626 | |||
| XM_011522621.2 | XP_011520923.1 | 5 | 5554 | |||
| XM_011522622.1 | XP_011520924.1 | 3 | 4258 | |||
| XM_011522623.2 | XP_011520925.1 | 6 | 2329 | |||
| XM_011522624.2 | XP_011520926.1 | 5 | 1667 | |||
| XM_011522625.2 | XP_011520927.1 | 4 | 1553 | |||
| XM_011522626.2 | XP_011520928.1 | 5 | 5118 | |||
| XM_011522627.2 | XP_011520929.1 | 5 | 5123 | |||
| XM_011522628.2 | XP_011520930.1 | 5 | 2187 | |||
| XM_011522629.2 | XP_011520931.1 | 5 | 2050 | |||
| XM_011522630.2 | XP_011520932.1 | 4 | 1388 | |||
| XM_011522631.2 | XP_011520933.1 | 3 | 1274 | |||
| XM_011522632.2 | XP_011520934.1 | 3 | 1705 | |||
| XM_017023596.1 | XP_016879085.1 | 6 | 2466 | |||
| GenBank mRNA | BC112918.1 | - | 3 | 1216 | ||
| U85647.1 | - | 4 | 1327 | |||

CAPN15
Genomic Map