siRNA Details
| Gene Symbol: CGRRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006568.2 | NP_006559.1 | 2 | 223 | ||
| XM_011536362.2 | XP_011534664.1 | 2 | 112 | |||
| XM_017020931.1 | XP_016876420.1 | 3 | 314 | |||
| XM_017020932.1 | XP_016876421.1 | 3 | 180 | |||
| GenBank mRNA | AK289478.1 | - | 2 | 254 | ||
| AK315318.1 | - | 2 | 264 | |||
| BC015063.1 | - | 2 | 189 | 4245551 | ||
| BG701924.1 | - | 2 | 284 | |||
| U66469.1 | - | 2 | 159 | |||

CGRRF1
Genomic Map