siRNA Details
| Gene Symbol: DTYMK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165031.1 | NP_001158503.1 | 2 | 327 | ||
| NM_001320902.1 | NP_001307831.1 | 2 | 327 | |||
| NM_001320903.1 | NP_001307832.1 | 2 | 327 | |||
| NM_001320904.1 | NP_001307833.1 | 2 | 327 | |||
| NM_001320905.1 | NP_001307834.1 | 2 | 327 | |||
| NM_012145.3 | NP_036277.2 | 2 | 327 | |||
| NR_033255.1 | 2 | 327 | ||||
| NR_135492.1 | 2 | 327 | ||||
| GenBank mRNA | AA427388.1 | - | 2 | 68 | ||
| AK223434.1 | - | 2 | 174 | |||
| BC001827.2 | - | 2 | 167 | 3533327 | ||
| BC147006.1 | - | 2 | 266 | 9056934 | ||
| BC147010.1 | - | 2 | 266 | 9056938 | ||
| BC171902.1 | - | 2 | 266 | 9054556 | ||
| BC171903.1 | - | 2 | 266 | |||
| BG765702.1 | - | 2 | 73 | |||
| BM552707.1 | - | 2 | 313 | |||
| CN356277.1 | - | 2 | 163 | |||
| CR541986.1 | - | 2 | 142 | |||
| CR542015.1 | - | 2 | 142 | |||
| CX166352.1 | - | 2 | 327 | |||
| L16991.1 | - | 2 | 184 | |||
| X54729.1 | - | 2 | 166 | |||

DTYMK
Genomic Map