siRNA Details
| Gene Symbol: YPEL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013313.4 | NP_037445.1 | 2, 3 | 497 | ||
| GenBank mRNA | AB098734.1 | - | 2, 3 | 497 | ||
| AB098735.1 | - | 2, 3 | 497 | |||
| AF060862.1 | - | 1, 2 | 197 | |||
| AF305069.1 | - | 1, 2 | 257 | |||
| BC034486.1 | - | 2, 3 | 355 | 4826997 | ||
| BC074501.1 | - | 2, 3 | 362 | 5171826 | ||
| BI461654.1 | - | 2, 3 | 551 | |||
| BI559478.1 | - | 2, 3 | 377 | |||
| CR456376.2 | - | 1, 2 | 144 | |||
| CR933630.1 | - | 1, 2 | 4600 | |||

YPEL1
Genomic Map