siRNA Details
| Gene Symbol: SLC16A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013356.2 | NP_037488.2 | 5 | 1359 | ||
| XM_017028685.1 | XP_016884174.1 | 4 | 1414 | |||
| GenBank mRNA | AF132610.1 | - | 5 | 1338 | ||
| BM714916.1 | - | 2 | 137 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC16A8| Gene Symbol: SLC16A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013356.2 | NP_037488.2 | 5 | 1359 | ||
| XM_017028685.1 | XP_016884174.1 | 4 | 1414 | |||
| GenBank mRNA | AF132610.1 | - | 5 | 1338 | ||
| BM714916.1 | - | 2 | 137 | |||