siRNA Details
| Gene Symbol: ZNF224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321645.1 | NP_001308574.1 | 4, 5 | 506 | ||
| NM_013398.3 | NP_037530.2 | 4, 5 | 464 | |||
| XM_017027261.1 | XP_016882750.1 | 3, 4 | 540 | |||
| GenBank mRNA | AF187990.2 | - | 4, 5 | 449 | ||
| AK292500.1 | - | 4, 5 | 444 | |||
| BC051902.1 | - | 4, 5 | 401 | 6155240 | ||
| CN304667.1 | - | 4, 5 | 330 | |||
| DA245440.1 | - | 4, 5 | 464 | |||
| DB228337.1 | - | 4, 5 | 496 | |||

ZNF224
Genomic Map