siRNA Details
| Gene Symbol: LOC102724229 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006716222.3 | XP_006716285.1 | 1 | 2026 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724229| Gene Symbol: LOC102724229 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006716222.3 | XP_006716285.1 | 1 | 2026 | ||