siRNA Details
| Gene Symbol: QPRT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014298.4 | NP_055113.2 | 2 | 369 | ||
| NR_134536.1 | 2 | 369 | ||||
| XM_005255223.3 | XP_005255280.2 | 3 | 452 | |||
| XM_017023101.1 | XP_016878590.1 | 3 | 452 | |||
| GenBank mRNA | BC005060.1 | - | 2 | 206 | 2960170 | |
| BC010033.2 | - | 2 | 198 | 3506460 | ||
| BC018910.2 | - | 2 | 223 | 3959973 | ||
| BG748194.1 | - | 2 | 339 | |||
| BJ997057.1 | - | 2 | 232 | |||
| BT007231.1 | - | 1 | 179 | |||
| D78177.1 | - | 1 | 179 | |||
| EU794638.1 | - | 2 | 198 | |||
| KJ534923.1 | - | 1 | 179 | |||

QPRT
Genomic Map