siRNA Details
| Gene Symbol: NKG7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005601.3 | NP_005592.1 | 2 | 356 | ||
| GenBank mRNA | BC015759.1 | - | 2 | 285 | 4849734 | |
| S69115.1 | - | 2 | 365 | |||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 NKG7
NKG7| Gene Symbol: NKG7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005601.3 | NP_005592.1 | 2 | 356 | ||
| GenBank mRNA | BC015759.1 | - | 2 | 285 | 4849734 | |
| S69115.1 | - | 2 | 365 | |||
 Back To Top
 Back To Top