siRNA Details
| Gene Symbol: GUCA2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007102.2 | NP_009033.1 | 2 | 149 | ||
| GenBank mRNA | BC069301.1 | - | 2 | 147 | 7262756 | |
| BC093779.1 | - | 2 | 143 | 7939624 | ||
| BC093781.1 | - | 2 | 142 | 7939626 | ||
| U34279.1 | - | 2 | 148 | |||
| Z50753.1 | - | 2 | 140 | |||

GUCA2B
Genomic Map