siRNA Details
| Gene Symbol: ZNF224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321645.1 | NP_001308574.1 | 5 | 518 | ||
| NM_013398.3 | NP_037530.2 | 5 | 476 | |||
| XM_017027261.1 | XP_016882750.1 | 4 | 552 | |||
| GenBank mRNA | AF187990.2 | - | 5 | 461 | ||
| AK292500.1 | - | 5 | 456 | |||
| BC051902.1 | - | 5 | 413 | 6155240 | ||
| CN304667.1 | - | 5 | 342 | |||
| DA245440.1 | - | 5 | 476 | |||
| DB228337.1 | - | 5 | 508 | |||

ZNF224
Genomic Map