siRNA Details
| Gene Symbol: PLLP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015993.2 | NP_057077.1 | 2 | 410 | ||
| GenBank mRNA | AF137386.1 | - | 2 | 406 | ||
| AK130584.1 | - | 2 | 402 | |||
| AK289599.1 | - | 2 | 387 | |||
| AK313910.1 | - | 2 | 410 | |||
| BC002760.2 | - | 2 | 373 | 3633345 | ||
| DA366260.1 | - | 2 | 411 | |||

PLLP
Genomic Map