siRNA Details
| Gene Symbol: GNG13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016541.2 | NP_057625.1 | 3 | 272 | ||
| GenBank mRNA | AB030207.1 | - | 3 | 251 | ||
| AF493880.1 | - | 2 | 170 | |||
| AK312139.1 | - | 3 | 273 | |||
| AY029486.1 | - | 3 | 238 | |||
| BC093760.1 | - | 2 | 203 | 7939605 | ||
| BC095525.1 | - | 2 | 173 | 6519181 | ||
| DA111448.1 | - | 3 | 273 | |||

GNG13
Genomic Map