siRNA Details
| Gene Symbol: TMEM160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017854.1 | NP_060324.1 | 2, 3 | 310 | ||
| XM_017026917.1 | XP_016882406.1 | 2, 3 | 605 | |||
| GenBank mRNA | AK000519.1 | - | 2, 3 | 310 | ||
| BC002907.2 | - | 2, 3 | 337 | 3941377 | ||

TMEM160
Genomic Map