siRNA Details
| Gene Symbol: FAM90A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319982.1 | NP_001306911.1 | 4 | 539 | ||
| NM_018088.3 | NP_060558.3 | 5 | 696 | |||
| XM_006719097.3 | XP_006719160.1 | 1 | 600 | |||
| XM_011520719.1 | XP_011519021.1 | 5 | 687 | |||
| XM_017019547.1 | XP_016875036.1 | 5 | 684 | |||
| GenBank mRNA | AK001270.1 | - | 4 | 522 | ||
| BC042608.1 | - | 5 | 675 | 5167203 | ||

FAM90A1
Genomic Map