siRNA Details
| Gene Symbol: CRCT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019060.2 | NP_061933.1 | 2 | 490 | ||
| XM_011509656.2 | XP_011507958.1 | 2 | 576 | |||
| GenBank mRNA | AJ243662.1 | - | 2 | 490 | ||
| BC119710.2 | - | 1 | 446 | 40117182 | ||

CRCT1
Genomic Map