siRNA Details
| Gene Symbol: ISOC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136201.1 | NP_001129673.1 | 2 | 257 | ||
| NM_001136202.1 | NP_001129674.1 | 2 | 257 | |||
| NM_024710.2 | NP_078986.1 | 2 | 257 | |||
| GenBank mRNA | AK027122.1 | - | 2 | 220 | ||
| AK131324.1 | - | 2 | 918 | |||
| AK311540.1 | - | 207 | ||||
| BC017344.2 | - | 2 | 144 | 4893552 | ||
| BP226043.1 | - | 2 | 258 | |||
| CR457377.1 | - | 1 | 83 | |||

ISOC2
Genomic Map