siRNA Details
| Gene Symbol: VPS28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF182844.1 | - | 907 | |||
| AF316887.1 | - | 889 | ||||
| AK026865.1 | - | 905 | ||||
| BC006485.1 | - | 891 | 3535787 | |||
| BC019321.1 | - | 891 | 2822758 | |||
| BC050713.2 | - | 1021 | 5741129 | |||

VPS28
Genomic Map