siRNA Details
| Gene Symbol: PBDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300888.1 | NP_001287817.1 | 3 | 353 | ||
| NM_016500.3 | NP_057584.2 | 3 | 346 | |||
| GenBank mRNA | BC001220.1 | - | 3 | 346 | 3528586 | |
| BC051894.1 | - | 3 | 208 | 6528202 | ||
| BM817702.1 | - | 3 | 242 | |||
| CD704296.1 | - | 3 | 247 | |||

PBDC1
Genomic Map