siRNA Details
| Gene Symbol: BLOC1S4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018366.2 | NP_060836.1 | 1 | 619 | ||
| GenBank mRNA | AF085849.1 | - | 1 | 12 | ||
| AK002092.1 | - | 1 | 619 | |||
| BC067815.1 | - | 1 | 560 | 30349339 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BLOC1S4| Gene Symbol: BLOC1S4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018366.2 | NP_060836.1 | 1 | 619 | ||
| GenBank mRNA | AF085849.1 | - | 1 | 12 | ||
| AK002092.1 | - | 1 | 619 | |||
| BC067815.1 | - | 1 | 560 | 30349339 | ||