siRNA Details
| Gene Symbol: LIN37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019104.2 | NP_061977.1 | 2 | 465 | ||
| GenBank mRNA | AK293116.1 | - | 2 | 230 | ||
| AK301992.1 | - | 2 | 465 | |||
| BC009071.1 | - | 2 | 184 | 3872616 | ||
| BI767008.1 | - | 2 | 215 | |||
| CB152701.1 | - | 2 | 267 | |||
| DC396639.1 | - | 2 | 465 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001290569, NM_029377, AK007858, BC013546, AY428767, and others.

LIN37
Genomic Map