siRNA Details
| Gene Symbol: ZNF117 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015852.3 | NP_056936.2 | 4 | 3861 | ||
| GenBank mRNA | M27879.1 | - | 4 | 1170 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF117| Gene Symbol: ZNF117 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015852.3 | NP_056936.2 | 4 | 3861 | ||
| GenBank mRNA | M27879.1 | - | 4 | 1170 | ||