siRNA Details
| Gene Symbol: CD276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024736.1 | NP_001019907.1 | 3 | 446 | ||
| NM_025240.2 | NP_079516.1 | 3 | 446 | |||
| XM_005254700.4 | XP_005254757.1 | 3 | 564 | |||
| XM_011522095.2 | XP_011520397.1 | 3 | 435 | |||
| XM_011522096.2 | XP_011520398.1 | 3 | 375 | |||
| XM_017022638.1 | XP_016878127.1 | 3 | 364 | |||
| GenBank mRNA | AF302102.1 | - | 2 | 212 | ||
| AJ583695.1 | - | 2 | 255 | |||
| AK074849.1 | - | 3 | 336 | |||
| AK075549.1 | - | 3 | 374 | |||
| AK291721.1 | - | 3 | 283 | |||
| AK292711.1 | - | 2 | 207 | |||
| AK296358.1 | - | 2, 3 | 164 | |||
| AK298522.1 | - | 3 | 344 | |||
| AK309728.1 | - | 3 | 283 | |||
| AY358343.1 | - | 3 | 363 | |||
| BC062581.1 | - | 3 | 446 | 5201491 | ||
| BQ278722.1 | - | 29 | ||||

CD276
Genomic Map