siRNA Details
| Gene Symbol: DEDD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270614.1 | NP_001257543.1 | 3 | 487 | ||
| NM_001270615.1 | NP_001257544.1 | 3 | 569 | |||
| NM_133328.3 | NP_579874.1 | 3 | 569 | |||
| NR_073046.1 | 2 | 203 | ||||
| NR_073047.1 | 2 | 121 | ||||
| NR_073048.1 | 2 | 203 | ||||
| NR_073049.1 | 2 | 203 | ||||
| NR_073050.1 | 2 | 203 | ||||
| NR_073051.1 | 2 | 203 | ||||
| NR_073052.1 | 2 | 203 | ||||
| XM_011526569.1 | XP_011524871.1 | 3 | 493 | |||
| XM_011526571.1 | XP_011524873.1 | 3 | 493 | |||
| XM_011526572.1 | XP_011524874.1 | 3 | 493 | |||
| XM_017026402.1 | XP_016881891.1 | 3 | 487 | |||
| XM_017026403.1 | XP_016881892.1 | 3 | 493 | |||
| GenBank mRNA | AF443591.1 | - | 2 | 370 | ||
| AF457575.1 | - | 2 | 370 | |||
| AK075328.1 | - | 3 | 493 | |||
| AK130203.1 | - | 2 | 111 | |||
| AK307895.1 | - | 3 | 461 | |||
| AY125488.1 | - | 3 | 453 | |||
| BC013372.2 | - | 3 | 444 | 3941907 | ||
| BC027930.1 | - | 3 | 438 | 5199772 | ||
| BM908148.1 | - | 2 | 115 | |||
| BX458265.2 | - | 2 | 153 | |||
| CN373297.1 | - | 2 | 160 | |||
| CR984238.1 | - | 2 | 118 | |||
| CT000394.1 | - | 2 | 117 | |||
| DA821401.1 | - | 3 | 488 | |||
| HY033141.1 | - | 2 | 124 | |||
| HY033487.1 | - | 2 | 127 | |||

DEDD2
Genomic Map