siRNA Details
| Gene Symbol: NEK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133494.2 | NP_598001.1 | 3 | 477 | ||
| XM_011509209.1 | XP_011507511.1 | 3 | 455 | |||
| XM_017000344.1 | XP_016855833.1 | 3 | 455 | |||
| XM_017000345.1 | XP_016855834.1 | 4 | 562 | |||
| XM_017000346.1 | XP_016855835.1 | 3 | 455 | |||
| XM_017000347.1 | XP_016855836.1 | 3 | 455 | |||
| XM_017000348.1 | XP_016855837.1 | 3 | 455 | |||
| GenBank mRNA | AB062450.1 | - | 2 | 196 | ||
| AK313411.1 | - | 3 | 277 | |||
| DB498485.1 | - | 3 | 477 | |||

NEK7
Genomic Map