siRNA Details
| Gene Symbol: KRTAP9-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031962.2 | NP_114168.1 | 1 | 570 | ||
| GenBank mRNA | AJ406947.1 | - | 1 | 570 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP9-3| Gene Symbol: KRTAP9-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031962.2 | NP_114168.1 | 1 | 570 | ||
| GenBank mRNA | AJ406947.1 | - | 1 | 570 | ||